If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Large Wiper Dispenser
- Dimensions:381×88.9×127 mm
- Cat. no.:102097-610
- Supplier No.:71602-L
Wipes holder fabricated of durable plastic will serve for year of use, yet are economical enough to have in every convenient spot in the lab.
- Instant mount with double sided tape
- Fit Kimwipes® and Accuwipe® brand wipes
- Available for small or large wipes boxes
Combo-Dispenser is also available which designed to hold most brand of wipes and gloves. Combo dispensers come with color white only.