Order Entry
Canada
ContactUsLinkComponent
Acrytol Mounting Medium, Electron Microscopy Science
Acrytol Mounting Medium, Electron Microscopy Science
Catalog # 102090-972
CAS Number:  
undefined
Acrytol Mounting Medium, Electron Microscopy Science
Catalog # 102090-972
Supplier Number:  13518
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Acrytol Mounting Medium
  • Size:
    1 pt
  • UN No.:
    1866
  • Boiling point:
    110 °C (230 °F)
  • Cat. no.:
    102090-972
  • Supplier No.:
    13518

Specifications

About this item

Acrytol is a rapid-drying mounting medium which enables slides to be screened immediately. It has a low viscosity, allowing the medium to flow easily. It also prevents air bubbles from becoming trapped.

Acrytol contains an antioxidant to inhibit stain fading and prevent the formulation of annual rings. A thin coating of Acrytol is all that is needed to adhere a coverglass to a microscope slide.