Contact Plate Polystyrene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Made of polystyrene. These dishes are free from optical distortion. The dish has 16 of 10 x 10 mm square grids, which is marked numerically in one direction and alphabetically in the other. The plate model with convex bottom is designed with this bottom to save on culture medium. Dishes are packaged 20 per sleeve and are radiation sterile.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together