Orders LinkContactUsLinkComponent
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Certificates
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Product Family Options
Product Information
  • Size
    Pack type
    Availability
    Price
  • 102092-216
    Phosphate Buffered Saline, 10X
    Phosphate Buffered Saline, 10X
    1 l
    Bottle
    Discontinued
    Supplier Number: 19342-10
  • 102092-218
    Phosphate Buffered Saline, 2X
    Phosphate Buffered Saline, 2X
    1 l
    Bottle
    Discontinued
    Supplier Number: 19343-10
  • 102092-222
    Phosphate Buffered Saline, 1X
    Phosphate Buffered Saline, 1X
    1 l
    Bottle
    Discontinued
    Supplier Number: 19344-10
  • 102092-210
    Phosphate Buffered Saline, 10X
    Phosphate Buffered Saline, 10X
    3.78 l
    Bottle
    Log in for availability
    Log in for availability
    Each
    Each - $333.24
    $333.24
    Each
       $333.24
    Supplier Number: 19242-40
  • 102092-220
    Phosphate Buffered Saline, 2X
    Phosphate Buffered Saline, 2X
    3.78 l
    Bottle
    Discontinued
    Supplier Number: 19343-40
  • 102092-224
    Phosphate Buffered Saline, 1X
    Phosphate Buffered Saline, 1X
    3.78 l
    Bottle
    Discontinued
    Supplier Number: 19344-20
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)