Rectangular Stain Dish, Electron Microscopy Sciences
Product Details & Documents
Heavy, clear, soda lime glass stain dish, accept glass slide measure 75 x 25 mm or 3 x 1”.
This dish comes with glass cover, holds 20 slides back-to-back horizontally.
Inside dimensions: 3 x 2 1/4x 1 1/4”.
Specifications
- Description:Rectangular Staining Dish
- Dimensions:3 x 2 1/4 x 1 1/4”
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)