Order Entry
Canada
ContactUsLinkComponent
Omnimount, Electron Microscopy Science
Omnimount, Electron Microscopy Science
Catalog # 102092-152
CAS Number:  
undefined
Omnimount, Electron Microscopy Science
Catalog # 102092-152
Supplier Number:  17997-01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Omnimount
  • Size:
    100 ml
  • Boiling point:
    162 °C (324 °F)
  • Cat. no.:
    102092-152
  • Supplier No.:
    17997-01

Specifications

About this item

Specially developed to provide compatibility with National Diagnostic clearing agents, HistoClear II. Omnimount™ combined outstanding optical characteristics with low fluororescence and exceptional durability.

  • Optical clarity combined with exceptional durability
  • Compatible with HistoClear II
  • Higher flash point and lower toxicity than xylene based mountants

In addition to being the ideal partner for HistoClear II, Omnimount is a truly universal mounting medium compatible with all common clearing agents: xylene, toluene, limonene, and petroleum derived products.

The Omnimount solvent has a higher flash point and a lower toxicity than xylene based mountants, so Omnimount™ provides both a safer work environment and reduce shipping costs.