AMS Sorbothane Balance Platform, Electron Microscopy Sciences
102090-930
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Ams sorbothane balance platform
- Environmentally Preferable:
- Cat. no.:102090-930
- Supplier No.:6870
The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7 Hz.
Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilization times and speeding up measurements.