ContactUsLinkComponent
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Catalog # 100502-820
Minority or Woman-Owned Business Enterprise
undefined
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Catalog # 100502-820
Supplier Number:  70175-50
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Stubs Thick Embedding 6 mm
  • Cat. no.:
    100502-820
  • Supplier No.:
    70175-50
Specifications
About this item
Block holders molded from hard plastic are easy to store as it attched to Aluminum chucks (ABH)
The large center hole in the holder prevents bubble formation and its frosted surface is easy for marking.