Orders LinkContactUsLinkComponent
PTFE Embedding Mold, Electron Microscopy Sciences
PTFE Embedding Mold, Electron Microscopy Sciences
Mold Set PTFE Flat Embedding
Catalog # 101454-042
Supplier #:  70143
PTFE Embedding Mold, Electron Microscopy Sciences
Catalog # 101454-042
PTFE Embedding Mold, Electron Microscopy Sciences
PTFE Embedding Mold, Electron Microscopy Sciences
Mold Set PTFE Flat Embedding
Price
$486.74
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For curing anaerobic material, we recommend using Aclar® film, cut-to-size of the mold and lower on top of the mold after you ʽjust over-fill’ the cavities with embedding resin
Machined from PTFE block. This mold offers flat embedding for  most resins, including LR White®, LR Gold®, JB-4, Immuno-Bed and Lowicryl®. This PTFE Flat Embedding Mold can be used for both UV/low temperature and thermal cure, and may be used as a substitution for the silicone molds. The individual cavities are pre shaped  to reduce block trimming.
For curing anaerobic material, we recommend using Aclar® film, cut-to-size of the mold and lower on top of the mold after you ʽjust over-fill’ the cavities with embedding resin. Due to the flexibility of the PTFE, you just bent the mold to pop out the cured resin blocks. After all the cast blocks have been removed, reinsert the mold to its holder to re-shape the mold. The mold holder is machined from aluminum and anodized black. Mold with 18 cavities, each cavity measures W x L x D : 5 x 14 x 4 mm. Overall mold measures: 140 x 50 x 8 mm (5 1/2 x 3 x 1/4”). Mold is available as a set (includes one PTFE mold and one aluminum holder).
Due to the flexibility of the PTFE, you just bent the mold to pop out the cured resin blocks. After all the cast blocks have been removed, reinsert the mold to its holder to re-shape the mold. The mold holder is machined from aluminum and anodized black. Mold with 18 cavities, each cavity measures W x L x D : 5 x 14 x 4 mm. Overall mold measures: 140 x 50 x 8 mm (5 1/2 x 3 x 1/4”). Mold is available as a set (includes one PTFE mold and one aluminum holder).
Specifications
  • Description:
    Mold Set PTFE Flat Embedding
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)