ContactUsLinkComponent
Polyethylene Molds, Individual Molds, Electron Microscopy Sciences
  101454-086
Minority or Woman-Owned Business Enterprise
undefined
Polyethylene Molds, Individual Molds, Electron Microscopy Sciences
  101454-086
 :  70177-19
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Mold Polyethylene
  • Size:
    19x13 mm
  • Cat. no.:
    101454-086
  • Supplier No.:
    70177-19
Molded from polyethylene, three popular sizes are available to meet your needs.
These Individual Molds can be used with EBH2 Block Holders, Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Comes with anodized blue and natural color. Suitable for JB-4®, JB-4-Plus®, Immuno-Bed, LR White, LR Gold® or Paraffin embedding.