Orders LinkContactUsLinkComponent
Adjustable Coverglass Rack, Electron Microscopy Sciences
Adjustable Coverglass Rack, Electron Microscopy Sciences
Adjustable Coverglass Rack, 48×48 mm (11/2×11/2")
Catalog # 101454-044
Supplier #:  72243
Adjustable Coverglass Rack, Electron Microscopy Sciences
Catalog # 101454-044
Adjustable Coverglass Rack, Electron Microscopy Sciences
Adjustable Coverglass Rack, Electron Microscopy Sciences
Adjustable Coverglass Rack, 48×48 mm (11/2×11/2")
Price
$499.11
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Innovative design saves you time and headache when you are looking for a cover-glass holder. This rack is machined from polypropylene and fabricated to accommodate a variety of cover-glasses in different sizes.
  • Holder is made from non ferrous material, it is microwaveable
With the ability to simply adjust the holder to fit your desired size and then securing  it by tightening  the reset screws with the hex key (hex key is included) all sizes of cover glasses are accommodated. The rack accommodates 10 cover glasses, round, square or rectangular, adjusting from 5 to 60 mm. Removable handle. Rack comes with two set of holding rods. One set accommodate cover glasses from 5 to 25 mm, and the other set for cover glasses from 5 to 60 mm.
Specifications
  • Description:
    Adjustable Coverglass Rack
  • Dimensions:
    48×48 mm (11/2×11/2")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)