If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Ultramicrotome Aid
- Cat. no.:102091-154
- Supplier No.:70612
Specifications
About this item
Ultramicrotome Aids are made from hard plastic (Delrin) with a beautifully machined finish to handle many of your instruments that need to be at your fingertips during sectioning.
This EMS Ultra-microtome Aid is a safe place to rest your diamond knife, diamond trimming tool, tweezers, hair curl or eye lash, diamond knife cleaning rods, perfect-loop for picking up sections.