If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Quadmag-Genie, 60 Hz
- Cat. no.:102098-016
- Electrical:120 V
- Supplier No.:72402-12
QuadMag Genie® is a general purpose multi-position magnetic stirrer designed for general laboratory applications which provides vigorous stirring for up to 4 vessels simultaneously.
The powerful magnet and motor offer exceptional magnetic force capable of stirring up to 2 L for each position. Powerful 'rare earth' magnets automatically pull stir bars into position and stay coupled even under the most demanding stirring conditions. Rugged ABS/Polycarbonate housing and the famous Vortex-Genie® heavy-duty motor provide years of reliable service. Spill-resistant housing channels fluids away from the internal components.