About this item
Goniometer Bases tightly capture the MicroRT™ tubing to allow seamless room-temperature and low-temperature data collection. Available in several different base styles that are compatible with MicroRT™ tubing and with standard cryo-tools, cryo-vials, magnetic goniometer head mounts, and most automating hardware.
- Base B1 is available in two versions: B1 (with flat ledge) and B1A (without the ledge, for ALS-style grippers used at ALS, NSLS and CHESS)
- Base B2 is a copper post-style base that accepts 11/12 mm MicroMounts™. Available only with the flat edge
- Base B3 is available in three versions: B3 for 19 mm MicroMounts™, with the flat ledge; B3A for 19 mm MicroMount™, without the ledge; and B3S for SPINE length MicroMount™, without ledge
- B4 is a 3 mm diameter “brass pin” style base, with the smaller central hole to tightly hold MicroMount™ and an extension to capture MicroRT™ tubing
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.