Order Entry
Canada
Orders LinkContactUsLinkComponent
Clear-Mount , Electron Microscopy Sciences

Clear-Mount , Electron Microscopy Sciences

Clear-Mount , Electron Microscopy Sciences
Certificates
Clear-Mount , Electron Microscopy Sciences
Clear-Mount , Electron Microscopy Sciences

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Application: Mounting of Immunohisto slides

Clear-Mount is a water based mounting medium designed for the permanent mounting of hydrated tissues, which may be damaged with organic solvents.  Such samples include cell smears, with peroxidase and alkaline phosphatase chromogens. This mounting medium preserves Fast Red, Aminoethylycarbazole (AEC), NBT/BCIP, INT/BCIP chromogens and is also compatible with counter-stain such as Hematoxylin and Nuclear Fast Red. It is also suitable for chromogens like DAB and DAB with Nickel and Cobalt. It is not compatible with H and E staining.

Application: Mounting of Immunohisto slides. Avalaible in original fomula with Tris Buffer or with PIPE Buffer for a more stable pH.

*The names BioMeda™ Gel Mount and Crystal Mount are all owned by BioMeda Corporation

*The names BioMeda™ Gel Mount and Crystal Mount are all owned by BioMeda Corporation

Avalaible in original fomula with Tris Buffer or with PIPE Buffer for a more stable pH.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100496-548
    Clear-Mount, (with Tris Buffer)
    Clear-Mount, (with Tris Buffer)
    250 ml
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $726.35
    $726.35
    Each (250ml)
       $726.35

    Supplier Number: 17985-15
  • 102092-130
    Clear-Mount (with PIPE Buffer)
    Clear-Mount (with PIPE Buffer)
    30 ml
    Discontinued
    Supplier Number: 17985-16
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 102092-132
    Clear-Mount (with PIPE Buffer)
    Clear-Mount (with PIPE Buffer)
    100 ml
    Discontinued
    Supplier Number: 17985-17
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.