ContactUsLinkComponent
Stepped Microtome Embedding Mold Dykstra, Electron Microscopy Sciences
Stepped Microtome Embedding Mold Dykstra, Electron Microscopy Sciences
Catalog # 102091-162
undefined
Stepped Microtome Embedding Mold Dykstra, Electron Microscopy Sciences
Catalog # 102091-162
Supplier Number:  70907
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Embedding Mold, Dykstra
  • Cat. no.:
    102091-162
  • Supplier No.:
    70907
Specifications
About this item
This mold produces blocks of the best dimension to be held securely by Reichert/Leica Ultracut stepped microtome chucks.
EMS Embedding Mold is a new mold designed with Michael J. Dykstra Ph.D. from North Carolina State University Department of Veterinary Medicine. There are 24 cavities and the depth of each cavity is 4 mm which will prevent breakage of the blocks during razor blade trimming. Mold Measurements: 85×77 mm (3³/₈×3"). Cavity Measurements: 15×7×4 mm.