Glassine Weighing Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Glassine is free of static electricity that plagues disposable plastic weighing dishes.
- No Static Build-Up
- Contaminant Free – Pure glassine is an ultra-clean paper product
- Preformed 3.5˝ diameter dish–convenient to use
- Color contrast – Chocolatecolored glassine reveals every particle of material
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)