Each complete CoolSafe System includes: 1 CoolSafe box with lid, 1 Cool Chamber and 2 Cool Bricks (1°C & –10°C).Chambers for 0.2 mL PCR plates or tubes, 384-well plates and flat-bottom microplates also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
The World’s Most Versatile and Effective Benchtop Cooling and Storage System
Versitile – Designed for 0.2ml tubes (strips or trays), 96- well and 384-well microplates, 0.5, 1.5, 5 mL tubes
Specially Formulated – Cool Bricks maintain a constant temperature (1°C or –10°C) for up to 10 hours
Quality Engineering – Each aluminum Cool Chamber surrounds tubes to guarantee sustained cold temperatures
Durable – The CoolSafe box is made of triple density expanded polystyrene and molded under extreme pressure creating a durable container that guarantees temperature stability.
Easy to Use – Easily interchange Cool Chambers and Cool Bricks for different sized tubes.
No Mess – Messy ice buckets are now a thing of the past.
Each complete CoolSafe System includes: 1 CoolSafe box with lid, 1 Cool Chamber and 2 Cool Bricks (1°C & –10°C).Chambers for 0.2 mL PCR plates or tubes, 384-well plates and flat-bottom microplates also include 4 holes for 1.5 mL tubes and 6 holes for 0.5 mL tubes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.