Order Entry
Canada
Orders LinkContactUsLinkComponent
Combination SEM Mount , Electron Microscopy Sciences

Combination SEM Mount , Electron Microscopy Sciences

Combo 45° and 90°, Aluminum
  101413-632
Supplier #:  75344
Combination SEM Mount , Electron Microscopy Sciences
  101413-632
Combination SEM Mount , Electron Microscopy Sciences
Combination SEM Mount , Electron Microscopy Sciences

Combo 45° and 90°, Aluminum

 
$54.39
   
Each
 
   
 

Aluminum slotted head with 3.1 mm diameter pin, fits most SEMs.

 
  • Description:
    Combo 45° and 90°
  • Material:
    Aluminum

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.