Orders LinkContactUsLinkComponent
Cork Stopper Vial Set, Electron Microscopy Sciences
Cork Stopper Vial Set, Electron Microscopy Sciences
Sample Storage Set, Sk-3
Catalog # 101412-918
Supplier #:  64269
Cork Stopper Vial Set, Electron Microscopy Sciences
Catalog # 101412-918
Cork Stopper Vial Set, Electron Microscopy Sciences
Cork Stopper Vial Set, Electron Microscopy Sciences
Sample Storage Set, Sk-3
Price
$358.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Most economical, straight-sided, flat bottom, glass shell vial with high quality cork closures. Ideally suited for storing small parts, geological specimens, soil and many other types of samples.
Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Vial size 3 dr (10.4 ml), 12 Ø×114 mm H, arrangement 5×20. Tray size 11½×3× 5".
Specifications
  • Description:
    Sample Storage Set, Sk-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.