General Purpose Transfer Pipet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
These pipettes are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipettes are bulk packed and the sterile are individually packed.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.