ContactUsLinkComponent
Large Dissecting Kit, Electron Microscopy Sciences
Large Dissecting Kit, Electron Microscopy Sciences
Catalog # 101413-594
Minority or Woman-Owned Business Enterprise
undefined
Large Dissecting Kit, Electron Microscopy Sciences
Catalog # 101413-594
Supplier Number:  72951-15
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Large Dissecting Kit, Kit
  • Cat. no.:
    101413-594
  • Supplier No.:
    72951-15
Specifications
About this item
This dissection kit includes the following:
  • 12 Scalpel Blades #11 with #3 Handle
  • 6 1/2" Kelly Hemostat, Straight
  • 6 ¼" Kelly Hemostat, Straight
  • 4 1/2" Iris Scissors
  • 51/2" Dissecting Scissors
  • Set of Dissecting Needle, one Straight and one Bent
  • 4 1/2" Straight Fine Point Serrated Jaws Forceps
  • 5 1/2" Straight Blunt tip Serrated Jaws Forceps
  • 2 mm Diameter Stainless Steel Punch
  • Suture Hook
  • Large Wood Handle Spatula EMS # Description Qty/Cs