Orders LinkContactUsLinkComponent
Cryogenic Labels, Electron Microscopy Sciences
Cryogenic Labels, Electron Microscopy Sciences
Cryogenic Labels, Electron Microscopy Sciences
Certificates
Cryogenic Labels, Electron Microscopy Sciences
Cryogenic Labels, Electron Microscopy Sciences
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cryogenic labels are made from top quality materials with wide range of sizes, designed specially for cryogenic use.
DuraStick Labels are suitable to be used under a temperature of −40 °C. These labels are features with smudge resistant surface and are available in rolls for thermal transfer printing or in 8½×11" sheets for laser printing. All accept hand written notations with cryo markers.
CryoStick Labels are suitable to be used for temperatures of −80 °C. Ideal for use with cryotubes, these labels are best used with thermal transfer printers or cryo marker pens. Available in a wide range of sizes on sheets or rolls, they are not recommended for laser printing.
LiquiStick Labels are suitable to be used for temperatures of −195 °C. Labels feature with the synthetic surface that accepts crisp bar codes from thermal transfer printers or handwritten notations cryo markers. Supplied in sheets or rolls, these labels are not recommended for use with laser printers.
Satin Cloth Labels are suitable to be used for temperatures of −60 °C. These textured labels withstand dry ice, deep freezers and liquid gases. Supplied in sheets or rolls. They are easily marked with cryo markers, not for use with automated printers.
Wrap-Around Labels are suitable for freezers and cryogenic use. These unique labels are for sub-zero use, measures ¹⁵/₁₆×2³/₈". They feature a ¹⁵/₁₆×¹⁵/₁₆" area, frosted for thermal printing or marking with cryo markers. The remainder of the label is clear. It wraps around the cryo-tube to provide a protective cover for printed information while leaving a clear space for viewing the tube contents.
Product Family Options
Product Information
  • Diameter
    Dimensions
    Packaging
    Availability
    Price
  • 101413-718
    Durastick labels
    Durastick labels
    0.5×0.9"
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $94.59
    $94.59
    Each
       $94.59
    Supplier Number: 77025-10
  • 101413-720
    Durastick labels
    Durastick labels
    0.5×1.75"
    500 labels/roll
    Discontinued
    Supplier Number: -
  • 101413-724
    Cryostick labels
    Cryostick labels
    7/16"
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $83.00
    $83.00
    Each
       $83.00
    Supplier Number: 77026-01
  • 101413-726
    Cryostick labels
    Cryostick labels
    0.5×0.9"
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $85.68
    $85.68
    Each
       $85.68
    Supplier Number: 77026-10
  • 101413-728
    Cryostick labels
    Cryostick labels
    0.5×1.5"
    500 labels/roll
    Discontinued
    Supplier Number: 77026-12
  • 102098-466
    Cryostick labels
    Cryostick labels
    0.5×1.75"
    1000 labels/roll
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $99.89
    $99.89
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $78.40
    $78.40
    Each
       $99.89
    Pack of 1,000
       $78.40
    Supplier Number: 77026-11
  • 101413-730
    Cryostick labels
    Cryostick labels
    0.75×0.9"
    500 labels/roll
    Discontinued
    Supplier Number: 77026-15
  • 101413-734
    Cryostick labels
    Cryostick labels
    0.75×1.5"
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $75.81
    $75.81
    Each
       $75.81
    Supplier Number: 77026-17
  • 101413-740
    Liquistick labels
    Liquistick labels
    0.5×1.75"
    500 labels/roll
    Discontinued
    Supplier Number: 77027-11
  • 101413-744
    Liquistick labels
    Liquistick labels
    0.75×0.9"
    500 labels/roll
    Log in for availability
    Log in for availability
    Each
    Each - $172.47
    $172.47
    Each
       $172.47
    Supplier Number: 77027-15
  • 102098-468
    Wrap-around labels
    Wrap-around labels
    0.94×2.375"
    1000 labels/roll
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $128.90
    $128.90
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $128.90
    $128.90
    Each
       $128.90
    Roll of 1,000
       $128.90
    Supplier Number: 77028-18
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)