ContactUsLinkComponent
Fortress™ FIB Sample Holder, Electron Microscopy Sciences
Fortress™ FIB Sample Holder, Electron Microscopy Sciences
Catalog # 101413-656
Minority or Woman-Owned Business Enterprise
undefined
Fortress™ FIB Sample Holder, Electron Microscopy Sciences
Catalog # 101413-656
Supplier Number:  75956-01
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    FIB Sample Holder
  • Cat. no.:
    101413-656
  • Supplier No.:
    75956-01
Specifications
About this item
Fortress™ FIB Holder with CastleGuard™ Protection are reusable holders that secure FIB sample that are held in a specific orientation without the use of adhesives, adhesive tabs, or conductive paints.
  • Unique clamping mechanism secures sample without over tightening
  • Standard 1/8" pin mount fits most FIBs and SEMs
  • Peripheral support structure protects specimens during handling and transport
  • Economical design offers user to prepare and store samples on the same holder
  • Compatible with Sample Sver™ Stoage containers for secure stoage and transport in an inert environment
Fortress™ FIB Holders can be used to position a thin, whole or cut TEM grid / disk in an orientation such that either in-situ or an ex-situ. FIB lift-out technique can be used to attach a FIB-prepared sample. Physical protection of the mounted sample on the grid is provided with the CastleGuard™ protection design. Up to 30 Fortress FIB Holders can be stored in a single SS200 Sample Saver™ storage container.
Fortress™ FIB Holder – Low Profile: Low Profile Fortress™ FIB Holder to hold either a standard FIB lift out specimen on a grid or an H-Bar cut Sample. Fortress™ Sample Loader Securely holds the Fortress™ Low Profile holder under a stereomicroscope and permits viewing from 2 angles without changing focus on the microscope.
Fortress™ FIB Holder – High Profile: High Profile Fortress™ FIB Holder to hold a standard FIB lift out specimen on a grid. Fortress™ Sample Loader securely holds the Fortress™ High Profile holder under a stereomicroscope and permits viewing from 2 angles without changing focus on the microscope.