Fluid Dispenser Bottles, Electron Microscopy Sciences
About this item
These ESD safe polyethylene solvent dispensers are available in three standard sizes.
- Surface resistivity of 10E7 to 10E9
These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated.
2 Options Available Below
Product Details & Documents
These ESD safe polyethylene solvent dispensers are available in three standard sizes.
- Surface resistivity of 10E7 to 10E9
These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated.
Product Family Options
Product Information
- AvailabilityPrice
- 101412-924Fluid Dispenser, 180 ml EaSupplier Number: 64299-02
- 101412-922Fluid Dispenser, 120 ml EaSupplier Number: 64299-01
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)