Orders LinkContactUsLinkComponent
British Standard Graticules - NG10, Electron Microscopy Sciences
British Standard Graticules - NG10, Electron Microscopy Sciences
Certificates
British Standard Graticules - NG10, Electron Microscopy Sciences
British Standard Graticules - NG10, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Originally designed by the National Coal Board for use in coal mining.
In this graticule the circle areas double progressively, hence the diameters alter by Root 2, so that the size classes can form a continuation of the standard series of sieves for particle sizing. Each particle is assigned to a size class defined by two adjacent circles, which represent the size limits of that class. Thus the distribution of size is obtained in terms of the diameter of circles having the same projected area as the particles. This method will cover particles in the range 150 micron to 0.38 micron. The size distributions with respect to their number and weight are determined separately. Final results are calculated as cumulative percents.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100499-242
    NG10 Graticule
    NG10 Graticule
    16 mm
    Log in for availability
    Log in for availability
    Each
    Each - 774,46 $
    774,46 $
    Each
       774,46 $
    Supplier Number: 68026-16
  • 100499-244
    NG10 Graticule
    NG10 Graticule
    19 mm
    Log in for availability
    Log in for availability
    Each
    Each - 774,46 $
    774,46 $
    Each
       774,46 $
    Supplier Number: 68026-19
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.