Order Entry
Canada
Orders LinkContactUsLinkComponent
Electrodes for Checker® pH Tester, Electron Microscopy Sciences

Electrodes for Checker® pH Tester, Electron Microscopy Sciences

Electrodes for Checker® pH Tester, Electron Microscopy Sciences
Certificates
Electrodes for Checker® pH Tester, Electron Microscopy Sciences
Electrodes for Checker® pH Tester, Electron Microscopy Sciences

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Replacement electrodes for Checker® tester.

The extended family of Checker® pH testers now includes standard gel-filled electrodes with built-in BNC or screw type connectors.

Product Family Options

Product Information

  • For use with
    Connector
    pH range
    Availability
    Price
  • 100493-014
    HI 1207 Replacement Electrode, Gel-Filled
    HI 1207 Replacement Electrode, Gel-Filled
    Checker 2 (legacy)
    Screw type
    0 - 14
    Discontinued
    Supplier Number: -
  • 100493-012
    HI 1271 replacement electrode
    HI 1271 replacement electrode
    Model 70699-02
    Screw type
    0 - 14
    Log in for availability
    Log in for availability
    Each
    Each - $57.34
    $57.34
    Each
       $57.34

    Supplier Number: 70700-01

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.