Orders LinkContactUsLinkComponent
Beaker Buddy™ Rack, Electron Microscopy Sciences
Beaker Buddy™ Rack, Electron Microscopy Sciences
16-place Beaker Buddy™
Catalog # 100493-762
Supplier #:  72365-01
Beaker Buddy™ Rack, Electron Microscopy Sciences
Catalog # 100493-762
Beaker Buddy™ Rack, Electron Microscopy Sciences
Beaker Buddy™ Rack, Electron Microscopy Sciences
16-place Beaker Buddy™
Price
$287.16
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Beaker Buddy™ racks are round, anodized aluminum rack for use with beakers, water baths and ice baths
Place the tubes in the rack, and then screw on the handle to hold down the cover. The handle is 51/8" long, and assist in the removal from hot beakers. Rack measures 3.5" diameter and holds up to 16 tubes.
These racks are ideal for use with 1.5 and 1.0 mL tubes. The rack has a thermometer slot for temperature monitoring and alimimnum legs for free-standing use.
Specifications
  • Description:
    16-place Beaker Buddy™
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)