Orders LinkContactUsLinkComponent
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
30 Slide Wing Handle Rack
Catalog # 100492-940
Supplier #:  70312-34
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
Catalog # 100492-940
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
30 Slide Wing Handle Rack
Price
$241.97
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This 30-slide rack is made from non-tarnishing stainless steel and it is resistant to most staining solutions.
The handle is permanently attached but it is swing-able to permitting for closure of the dish and easy insertion and removal of the slides.
Specifications
  • Description:
    30 Slide Wing Handle Rack
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.