ContactUsLinkComponent
BD* Carbon Steel Scalpel Blade 60, Electron Microscopy Sciences
BD* Carbon Steel Scalpel Blade 60, Electron Microscopy Sciences
Catalog # 100500-134
Minority or Woman-Owned Business Enterprise
undefined
BD* Carbon Steel Scalpel Blade 60, Electron Microscopy Sciences
Catalog # 100500-134
Supplier Number:  72050-10
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    60 Scalpel Blade
  • Cat. no.:
    100500-134
  • Supplier No.:
    72050-10
Specifications
About this item
Scalpel blades made from surgical carbon steel, size 60 with convex cutting edge ideal for histology labs. 2 3/8" (6 cm) long and ribbed for extra rigidity. Non-sterile and are wrapped individually. Bard-Parker brand.