Gold Seal Fluorescent Antibody Slides, Electron Microscopy Sciences
100498-560
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Capacity:25 g
- Description:Fluorescent antibody slides
- L×W:75×25 mm (3×1")
- Environmentally Preferable:
- Cat. no.:100498-560
- Supplier No.:63732-02
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.