Ultra Thin Light Box - Light Pad, Electron Microscopy Sciences
Product Details & Documents
Very thin - only ½" thick (14 mm). Modern technology provides extremely even, bright illumination. The unit can be used flat on a table, standing at an incline, or hung on a wall.
- viewing surface: 8¾×11½" (22×29.2 cm)
- Two 5400 K color corrected lamps with 20000 hour life
- The overall size is 12¼×9¼×½"
- Weighs 2 lbs.
The light pad is also designed to display photo transparencies held under a clear plastic panel. Fits in a briefcase or stores in a desk drawer. It is powered by a 12 V, DC adapter.
Specifications
- Description:Ultra Thin Light Box
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)