ContactUsLinkComponent
Wood Applicators, Electron Microscopy Sciences
Wood Applicators, Electron Microscopy Sciences
  100501-816
Minority or Woman-Owned Business Enterprise
undefined
Wood Applicators, Electron Microscopy Sciences
  100501-816
 :  72304-10
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Double Bevel Applicator Stick
  • Dimensions:
    7 x 5/32"
  • Cat. no.:
    100501-816
  • Supplier No.:
    72304-10
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring, and mixing.