V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
About this item
V-Vials are ideal for: micro mixing, homogenization, centrifugation, concentration, extraction, derivatization, radiosynthesis, hybridization, anaerobic reactions, and catalytic hydrogenation.
- All components can be autoclaved
- Graduated and clear
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
Screw cap has solid top and TFE-faced has white (styrene butadiene) rubber liner.
Packaging: They come packed 12 vials in a hinged lid storage box.
5 Options Available Below
Product Details & Documents
V-Vials are ideal for: micro mixing, homogenization, centrifugation, concentration, extraction, derivatization, radiosynthesis, hybridization, anaerobic reactions, and catalytic hydrogenation.
- All components can be autoclaved
- Graduated and clear
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
Screw cap has solid top and TFE-faced has white (styrene butadiene) rubber liner.
Packaging: They come packed 12 vials in a hinged lid storage box.
Product Family Options
Product Information
- CapacityCap includedAvailabilityPrice
- 0.30 mlYesEach$473.47Pack of 12$332.59Supplier Number: 72647-01
- 1.00 mlYesEach$534.74Pack of 12$385.20Supplier Number: 72647-02
- 3.00 mlYesEach$370.52Pack of 12$519.60Supplier Number: 72647-07
- 5.00 mlYesEach$724.07Pack of 12$505.95Supplier Number: 72647-08
- 2.00 mlYesEach$646.03Pack of 12$463.26Supplier Number: 72647-05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.