Shell Vials, Electron Microscopy Sciences
About this item
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage.
Delivery information: The vials come with polyethylene open bottom snap-caps, Titeseal®, which are packed separately. Except the 4- dram size vial which comes without a closure. We recommend the use of our cork stoppers.
6 Options Available Below
Product Details & Documents
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage.
Delivery information: The vials come with polyethylene open bottom snap-caps, Titeseal®, which are packed separately. Except the 4- dram size vial which comes without a closure. We recommend the use of our cork stoppers.
Product Family Options
Product Information
- Cap includedAvailabilityPrice
- 100489-506Shell vials, 1 DramShell vials, 1 DramYesEach$179.67Pack of 144$136.56Supplier Number: 72631-10
- 100502-744Shell vials, 2 DramShell vials, 2 DramYesEach$342.03Pack of 144$260.35Supplier Number: 72631-20
- 100489-508Shell vials, 3 DramShell vials, 3 DramYesEach$227.91Pack of 144$191.62Supplier Number: 72631-30
- 100489-498Shell vials, 0.5 DramShell vials, 0.5 DramYesEach$176.49Pack of 144$152.88Supplier Number: 72630-05
- 100489-510Shell vials, 4 DramShell vials, 4 DramNoEach$132.89Pack of 144$101.28Supplier Number: 72631-40
- 100489-512Shell vials, 7 DramShell vials, 7 DramYesEach$500.41Pack of 144$381.89Supplier Number: 72631-70
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)