Order Entry
Canada
Orders LinkContactUsLinkComponent
Vernier Caliper, Electron Microscopy Sciences

Vernier Caliper, Electron Microscopy Sciences

Vernier Caliper
Catalog # 100496-132
Supplier #:  62064
Vernier Caliper, Electron Microscopy Sciences
Catalog # 100496-132
Vernier Caliper, Electron Microscopy Sciences
Vernier Caliper, Electron Microscopy Sciences

Vernier Caliper

Price
$46.14
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Double scale vernier caliper. Readable on the vernier scale to 1/128" and/or to 0.1 mm.

  • Nickel plated scale
  • Roller adjustment

Inch Scale: Reading from 0 to 5" in 1/16" divisions
Metric Scale: Reading from 0 to 120 mm in 1 mm divisions

 : Packed in a plastic case.

Specifications
  • Description:
    Vernier Caliper

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.