Specifications
- Description:Vacuum Bell Jar & Plate
- Supplier No.:71236
Specifications
About this item
Good for vacuum drying and degassing procedures.
The chamber is made from a clear polycarbonate allowing for full visibility. Its polypropylene stopcock has a "PTFE" plug and it accepts 6.4 mm (1/4") I.D. tubing. The chamber measures: 20 cm (7 7/8") I.D.; inside maximum height is 11 cm (4 3/8"); plate is 4.4 cm (1 3/4") x 23.2 cm (9 1/8") in diameter.