ContactUsLinkComponent
The Mold for Long Tissues, Electron Microscopy Sciences
The Mold for Long Tissues, Electron Microscopy Sciences
Catalog # 100491-604
Minority or Woman-Owned Business Enterprise
undefined
The Mold for Long Tissues, Electron Microscopy Sciences
Catalog # 100491-604
Supplier Number:  70166
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mold for Long Tissues
  • Cat. no.:
    100491-604
  • Supplier No.:
    70166
Specifications
About this item
This mold accommodates longer pieces of tissues and orient them properly
The tissues lie flat in the well therefore no horizontal trimming is needed. It is ideal for any specimen that needs immediate sectioning (e.g. cell monolayer). The mold has 24 cavities in it.