ContactUsLinkComponent
The England Finder, Electron Microscopy Sciences
The England Finder, Electron Microscopy Sciences
  100499-390
Minority or Woman-Owned Business Enterprise
undefined
The England Finder, Electron Microscopy Sciences
  100499-390
 :  68048-07
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    S7 England Finder Slide
  • Cat. no.:
    100499-390
  • Supplier No.:
    68048-07
S7 is a glass slide 75 x 26 mm (3 x 1") marked over the top surface in a way that a referenced position can be directly read relative to the locating edges. That means it is marked with a square grid at 1mm intervals. Each square contains a center ring bearing reference letters and numbers, the remainder of the square being subdivided into four segments numbered 1 to 4.
  • Reference numbers run horizontally 1 to 75 and letters vertically A to Z (omitting I)
  • Main locating edge is the bottom of the slide, which is used in conjunction with either the left or right vertical edge of the slide
  • A total of 100 positions on a slide can be accurately located
All England Finders produced for over 40 years are identical. The purpose of the finder is to give the microscopist an easy method of recording the position of a particular field of interest, so that another person in another laboratory can relocate at a later date, or the same position, or when using any other England Finder on any other microscope.
The location of the arrows is identical for all England Finder slides. Mark a label on the left hand side of the specimen slide, indicating the orientation to be repeated. By replacing the specimen slide with England Finder, taking care not to disturb the position, feature of interest can be noted. The feature can also be re-located at another place or time by reversing the procedure. A total of 100 positions on a slide can be accurately located.