Orders LinkContactUsLinkComponent
TEM Checker™, Electron Microscopy Sciences
Tem Checker
Catalog # 100492-374
 :  80059
TEM Checker™, Electron Microscopy Sciences
Catalog # 100492-374
CAS Number:  
TEM Checker™, Electron Microscopy Sciences
TEM Checker™, Electron Microscopy Sciences
Tem Checker
CAS Number:  
Price
$1,376.26
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TEM Checker contains 5 manganese discs in a standard grid storage box. The discs are not transparent to the beam but gives a strong manganese peak to check the resolution of EDS detector.
  • Monitor the performance of their x-ray detectors
  • Each disc is 3 mm in diameter and fits into the sample holder
Specifications
Specifications
  • Description:
    Tem Checker
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)