PTFE Beakers, Electron Microscopy Sciences
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
- With pouring spout
- Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
- With pouring spout
- Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.
Product Family Options
- CapacityDescriptionColorHeightExt.Ø
- : 60936
- : 60938
- : 60940
- : 60941
- : 60942
- : 60947
- : 60946
- 500 mlPTFE BeakersWhite125 mm73 mm-: 60948
- 600 mlPTFE BeakerWhite127 mm80 mm-: 60949-06
- : 60949-10
- : 60949-20
- : 60936-C
- : 60938-C
- : 60940-C
- : 60942-C
- : 60944-C
- : 60946-C
- : 60947-C
- : 60948-C
- : 60949-06C
- 1000 mlPTFE Beaker CoverWhite-: 60949-10C
- : 60941-C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)