Orders LinkContactUsLinkComponent
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Certificates
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The forceps are made from stainless steel and have medium points, serrated tips and alignment pin.
  • With curved or straight tips
  • Available in 4" and 6" long
  • Curved type A tips comes with locking device
Product Family Options
Product Information
  • Material
    Length
    Tip configuration
    Availability
    Price
  • 100493-956
    Forceps, 2×1 mm Slot
    Forceps, 2×1 mm Slot
    Stainless steel
    4"
    Straight Tips
    Log in for availability
    Log in for availability
    Each
    Each - $45.05
    $45.05
    Each
       $45.05
    Supplier Number: 72991-4S
  • 100493-954
    Forceps, 2×1 mm Slot
    Forceps, 2×1 mm Slot
    Stainless steel
    4"
    Curved Tips
    Log in for availability
    Log in for availability
    Each
    Each - $52.42
    $52.42
    Each
       $52.42
    Supplier Number: 72991-4C
  • 100493-958
    Forceps, 2×1 mm Slot
    Forceps, 2×1 mm Slot
    Stainless steel
    6"
    Curved "A" Tips
    Log in for availability
    Log in for availability
    Each
    Each - $45.05
    $45.05
    Each
       $45.05
    Supplier Number: 72992-6C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)