Orders LinkContactUsLinkComponent
Staining Jar, Electron Microscopy Sciences
Staining Jar, Electron Microscopy Sciences
Staining Jar with Ground Lid
Catalog # 100490-750
Supplier #:  70316-02
Staining Jar, Electron Microscopy Sciences
Catalog # 100490-750
Staining Jar, Electron Microscopy Sciences
Staining Jar, Electron Microscopy Sciences
Staining Jar with Ground Lid
Price
$640.02
Log in for availability
Each Pack of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An all glass jar which holds 5 to 10 standard slides (3 x 1").
They can be used as a chromatography jar with a choice of 2 ground glass lids. The lids are ground to the flange of the jar which deters evaporation. The jar with a taller lid accommodates larger slides: 100 x 25 mm (4 x 1").
Specifications
  • Description:
    Staining Jar with Ground Lid
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)