Orders LinkContactUsLinkComponent
Stabilizing Rings, Electron Microscopy Sciences | Avantor
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Rings, Electron Microscopy Sciences
Certificates
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Rings, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Product Family Options
Product Information
  • Availability
    Price
  • 100501-586
    Stabilizing Ring, 48 mm id
    Stabilizing Ring, 48 mm id
    Log in for availability
    Log in for availability
    Each
    Each - $93.89
    $93.89
    Each
       $93.89
    Supplier Number: 64436
  • 100496-214
    Stabilizing Ring, 20 mm id
    Stabilizing Ring, 20 mm id
    Log in for availability
    Log in for availability
    Each
    Each - $99.97
    $99.97
    Each
       $99.97
    Supplier Number: 64435
  • 100489-790
    Stabilizing Ring, Assorted Sizes
    Stabilizing Ring, Assorted Sizes
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $420.75
    $420.75
    Log in for availability
    Pack of 5
    Pack of 5 - $302.57
    $302.57
    Each
       $420.75
    Pack of 5
       $302.57
    Supplier Number: 64440
  • 100496-218
    Stabilizing Ring, 71 mm id
    Stabilizing Ring, 71 mm id
    Log in for availability
    Log in for availability
    Each
    Each - $148.46
    $148.46
    Each
       $148.46
    Supplier Number: 64439
  • 100502-372
    Stabilizing Ring, 56 mm id
    Stabilizing Ring, 56 mm id
    Log in for availability
    Log in for availability
    Each
    Each - $88.65
    $88.65
    Each
       $88.65
    Supplier Number: 64437
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.