If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Cabinet System Base
- Cat. no.:100501-474
- Supplier No.:74780-10
Specifications
About this item
These cabinets for filing 3x1" slides is the most effective system available to the ever-growing problem of slide storage and retrieval.
- Store up to 54,000 slides in only 2 square feet of space
- Low cost slide storage
- Sturdy - Built to last
The cabinets are stackable and are secured by interlocks. The base (order separately) lifts the cabinets 4¾"" off the floor. All units are constructed from heavy gauge reinforced cold rolled steel, electrostatically finished in dura-baked enamel in an attractive green color. Drawers are easily removable and interchangeable. They have rolled edges on each side for added strength. The riveted card holder on the front of the drawer speeds up slide retrieval. Each cabinet has 6 drawers, with two compartments per row, so you can store as many as 750 slides per drawer. The outside walls and center divider are lower than the slides, allowing for easy removal of any slide. Each drawer has an exclusive built-in hand grip at the rear and a full sized handle at the front which allows the drawers to be removed and used as trays. Each cabinet comes with a package of Memorandum Markers and a set of movable Compressor Blocks. The cabinet measures: 15¾"(W) x 18¾"(D)x4¾" (H) (40 x 47 x 12 cm).
Ideal for use in Pathology, Cytology, Hematology, Histology, Dermatology, Research, Education.