Slide Staining Dish, Electron Microscopy Sciences
The dish is made from polymethylpentene "glass clear" for use with our Polypropylene staining rack.
It comes with two lids; one lid should be used when the rack is not inserted.
Dimensions: 81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8").
- Description:Staining Dish
- Dimensions:81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)