SEM Sample Holder Set , Electron Microscopy Sciences
Catalog # 100492-278
Supplier: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:SEM Sample Holder Set
- Cat. no.:100492-278
- Supplier No.:75930
Specifications
About this item
For your convenience we now carry a SEM sample holder set to fill all of your needs.
Ordering information: Please specify the base requirement for your instrument when ordering. Delivery time on this item is approximately 2 weeks.
Delivery: The set consists of one universal holder, one vertical mount holder for thin samples, one 3-pin and one 5-pin configuration holder with a key for the set screw all in a finely finished wood box.