ContactUsLinkComponent
Slide Duplicating With Cover Slip Mold, Electron Microscopy Sciences
Catalogue # 100491-610
Minority or Woman-Owned Business Enterprise
undefined
Slide Duplicating With Cover Slip Mold, Electron Microscopy Sciences
Catalogue # 100491-610
Supplier Number:  70172
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Slide Duplicating withCover Slip
  • Cat. no.:
    100491-610
  • Supplier No.:
    70172
Spécifications
À propos de cet article
The same design as slide duplicating mold, but with the addition of three round cavities.
Cavities are 17 mm in diameter and 1 mm deep.