If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Slide Duplicating Mold
- Cat. no.:100491-608
- Supplier No.:70170
Specifications
About this item
It uses a 1×3" glass slide as a base, and duplicates the slide using the resin of your choice.
Tissue sections are embedded against the flat glass surface. This duplicated resin slide can be studied under the light microscope, then cut, remounted, resectioned, and studied under the EM. A three cavity mold, each cavity measuring: 1W×3³/₃L×2"D. Reference instructions enclosed.