ContactUsLinkComponent
Sapphire Burnishing/Deburring Tool, Electron Microscopy Sciences
Sapphire Burnishing/Deburring Tool, Electron Microscopy Sciences
Catalog #: 100497-994
Minority or Woman-Owned Business Enterprise
SAPPHIRE BURNISHING TOOL
Sapphire Burnishing/Deburring Tool, Electron Microscopy Sciences
Catalog #: 100497-994
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Sapphire Burnishing Tool
  • Supplier No.:
    62160-00
Specifications
About this item
This tool will out-perform any stone for the fine burnishing of soft or hard materials such as; miniature pivots, shafts, and pins that are made from hard steel, carbide and even ceramic. This precisely ground sapphire blade is 1.5×2.3 mm, 14 mm long (0.06x0.09x.56"). It is permanently mounted in a hexagonal gold anodized aluminum handle. Total length is 120 mm (4.75").
Customers Who Bought This Also Bought